Learn More
MOLTOX Molecular Toxicology Inc Top Agar w/ 0.6% Histidine/Biotin, 500 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948749
Encompass
For use with Minimal Glucose Agar and S. typhimurium strains in the Ames assay. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs.
Catalog No. 50-948-749 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26545
For use with Minimal Glucose Agar and S. typhimurium strains in the Ames assay. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs.