Learn More
MOLTOX Molecular Toxicology Inc Top Agar w/ L-Histidine & D-Biotin, 500 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948730
Encompass
For use with Minimal Glucose Agar and S. typhimurium strains in the Ames assay. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.
Catalog No. 50-948-730 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26503.5
For use with Minimal Glucose Agar and S. typhimurium strains in the Ames assay. MOLTOX provides all products necessary to conduct the Salmonella, E. coli, MOLTOX FT™, µAmes and UMU Mutagenicity Assays. All products are formulated and manufactured using the highest quality components and are consistent with the recommendations set forth by the historical literature corresponding to each test type. Certain Ames materials (e.g. STDiscs™ and Lyophilized S9) have been specifically developed in our laboratory for these assays. All products are thoroughly tested for performance in the assay and are accompanied by Quality Control and Formulation statements.