Learn More
SP Hotpack™ Top and Bottom Rack Set
Catalog No. 15365002 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Holds 22 spindles per rack
Catalog No.
15-365-002 Holds 22 spindles per rack
1 options
Catalog No. 15-365-002 Supplier SP Hotpack™ Supplier No. 437080
Useful for cleaning inside narrow-neck glassware, these spindle racks are designed for use with undercounter, freestanding and Vertical Stacking SP Hotpack™ Glassware Washer models.
  • Top and bottom spindle rack set for SP Hotpack glassware washers; bottom full rack with 22 spindles, top full rack with 22 spindles, replaces top and bottom open racks, Hotpack spindle rack ready washers only
  • Recommended for graduated cylinders, volumetric flasks, round bottom flasks, Erlenmeyer flasks and other narrow-neck glassware
  • Multiple configurations available to suit a variety of glassware
Note: Spindle racks fit only in units that are spindle rack ready.
Specifications
Description Holds 22 spindles per rack (2 racks)
Product Type Glassware Washer Accessory
For Use With (Application) Cleaning inside narrow-neck glassware
For Use With (Equipment) Hotpack Spindle Rack Ready Washers Only