Learn More
Mettler Toledo™ Top Cover Pole EasyPlace
Catalog No. 01911944 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-944 EasyPlace
1 options
This item is not returnable. View return policy
Catalog No. 01-911-944 Supplier Mettler Toledo™ Supplier No. 30671581
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cover for the top of the EasyPlace pole. Contains two positions for long term sensor storage.
  • Accessory
TRUSTED_SUSTAINABILITY
Specifications
Type Top Cover Pole EasyPlace
For Use With (Equipment) EasyPlace