Learn More
Ampco Top Cutting Pliers
SDP
Catalog No. 19173521 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
7 -1/4 in.
Catalog No. Length (English)
19-173-521 7 -1/4 in.
1 options
Catalog No. 19-173-521 Supplier Ampco Supplier No. 8350
Add to Cart
Add to Cart
Precision, non-sparking, non-magnetic cutting tools designed for use where ordinary steel pliers could ignite flammable atmospheres or interfere with magnetic equipment.
  • Made in Germany
  • Length of 185 mm or 7 and 1/4 in.
Specifications
Product Type Top Cutting Pliers
Length (English) 7 -1/4 in.
Length (Metric) 185 mm
No. per Pack 1 Ea.