Learn More
Leica Biosystems Top DM6 B 5-fold fluorescence turret (for Leica DM6 B microscopes)
Catalog No. NC3365967
Encompass_Preferred
Top DM6 B 5-fold fluorescence turret (MPN 11889202) is a motorized top module for the Leica DM6 B upright microscope that provides five selectable fluorescence cube positions for multi-channel imaging. It accepts fluorescence filter cubes and enables rapid channel switching for routine and advanced fluorescence workflows.
  • Motorized five-position cube turret.
  • Designed for fluorescence cube/filter based channel switching.
  • Fits DM6 B upright microscope top module interface.
  • Enables rapid, repeatable switching between filter cubes.
  • Intended for multi-channel fluorescence imaging applications.
Catalog No. NC3365967 Supplier Leica Biosystems Supplier No. 11889202
Add to Cart
Add to Cart
Request a Quote
Top DM6 B 5-fold fluorescence turret (MPN 11889202) is a motorized top module for the Leica DM6 B upright microscope that provides five selectable fluorescence cube positions for multi-channel imaging. It accepts fluorescence filter cubes and enables rapid channel switching for routine and advanced fluorescence workflows.
  • Motorized five-position cube turret.
  • Designed for fluorescence cube/filter based channel switching.
  • Fits DM6 B upright microscope top module interface.
  • Enables rapid, repeatable switching between filter cubes.
  • Intended for multi-channel fluorescence imaging applications.