Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Top Dog 6 mil Large Gown

Catalog No. 17227969 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-969 Red
17-227-971 Yellow
17-227-967 Green
17-227-968 Blue
17-227-970 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-227-969 Supplier Vikan™ Supplier No. 86034
Add to Cart
Add to Cart
This item is not returnable. View return policy

Size large open-back gowns feature full-length cuffed sleeves and are available in a variety of colors.

  • Combines the protection of sleeves and aprons into one garment
  • Open back improves user comfort by reducing weight and providing ventilation
  • Reusable
  • Color-coded
  • FDA CFR 21 compliant

Specifications

Style Full Length
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Length (English) 45 in.
For Use With (Application) Food Processing, Handling, Service
Length (Metric) 114.3 cm
Size Large
Material Ether-Based Polyurethane
Color Red
Closure Type Over-the-Head
Disposable Reusable
Sterility Non-sterile
Cuff Style Elastic
Model 86034
Sleeve Style Long Sleeve
Thickness 6 mil
Certifications/Compliance FDA 21 CFR
Autoclavable Not Autoclavable
No. per Pack 10/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.