Learn More
Felix Storch Inc ACCUCOLD Top Freezer Refrigerator 30 inch Wide
SDP
Catalog No. NC3901252
Encompass_Preferred
SUMMIT's selection of top freezer refrigerators offers true convenience for every kitchen, including break rooms and other general purpose facilities.The CTR18PL is a full-sized refrigerator-freezer. The large interior offers 18 cu.ft. of storage capacity, with frost-free operation for low maintenance use. It comes in a textured gray cabinet with user-reversible fingerprint-free doors that offer the look of stainless steel without the constant cleanup.Inside, the CTR18PL includes adjustable glass shelves in the refrigerator compartment and two clear crispers. The door features full-sized storage racks, including one that holds full gallon containers. Two half-sized door racks are adjustable to accommodate different sized items, with an additional fixed butter dish. The upper freezer compartment comes with a full-width wire shelf and two door bins. This unit includes an automatic interior LED light and easy-to-use interior thermostat: https://www.accucold.com/product/CTR18PL
Catalog No. NC3901252 Supplier Felix Storch Inc Supplier No. CTR18PL
Add to Cart
Add to Cart
SUMMIT's selection of top freezer refrigerators offers true convenience for every kitchen, including break rooms and other general purpose facilities.The CTR18PL is a full-sized refrigerator-freezer. The large interior offers 18 cu.ft. of storage capacity, with frost-free operation for low maintenance use. It comes in a textured gray cabinet with user-reversible fingerprint-free doors that offer the look of stainless steel without the constant cleanup.Inside, the CTR18PL includes adjustable glass shelves in the refrigerator compartment and two clear crispers. The door features full-sized storage racks, including one that holds full gallon containers. Two half-sized door racks are adjustable to accommodate different sized items, with an additional fixed butter dish. The upper freezer compartment comes with a full-width wire shelf and two door bins. This unit includes an automatic interior LED light and easy-to-use interior thermostat: https://www.accucold.com/product/CTR18PL