Learn More
MSC Milwaukee Tool Saw Chain, 14 in, .325 in Low Profile Pitch, .043 in Gauge
Catalog No. 502835532
Encompass
This 14 in .325 in Low Profile Pitch, .043 in Gauge Saw Chain delivers fast hardwood cutting with a multi-axis shape grind and is designed to prevent stalls in sawing applications. Its narrow kerf, low profile cutters offer more cuts per charge versus standard kerf to help maximize battery efficiency.
  • Delivers fast hardwood cutting with multi-axis shape grind.
  • Designed to prevent stalls during sawing.
  • Features narrow kerf, low profile cutters for increased cuts per charge.
  • Provides low kickback.
  • Has a standard chain sequence.
  • Requires a 5/32 inch file size.
  • Uses a 25 degree file angle.
  • Has a .025 inch depth gauge.
Catalog No. 50-283-5532 Supplier MSC Supplier No. 51887495
This 14 in .325 in Low Profile Pitch, .043 in Gauge Saw Chain delivers fast hardwood cutting with a multi-axis shape grind and is designed to prevent stalls in sawing applications. Its narrow kerf, low profile cutters offer more cuts per charge versus standard kerf to help maximize battery efficiency.
  • Delivers fast hardwood cutting with multi-axis shape grind.
  • Designed to prevent stalls during sawing.
  • Features narrow kerf, low profile cutters for increased cuts per charge.
  • Provides low kickback.
  • Has a standard chain sequence.
  • Requires a 5/32 inch file size.
  • Uses a 25 degree file angle.
  • Has a .025 inch depth gauge.