Learn More
MSC AbilityOne Floor Pad, Tan Burnishing, 14 in Diameter, 3400 Series
Catalog No. 502839664
Encompass
This tan burnishing floor pad is engineered for optimal performance with high-speed rotary disk floor machines exceeding 1500 rpm. It effectively cleans and polishes a variety of surfaces, including resilient tile, terrazzo, marble, and ceramic, consistently producing a "wet look" gloss in high-traffic environments.
  • Designed for rotary disk floor machines operating above 1500 rpm
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors
  • Can be washed and reused for extended life
  • Corrosion-resistant materials ensure durability
  • Produces a high-gloss "wet look" finish
  • Features an 11/16 in thickness for consistent performance
Catalog No. 50-283-9664 Supplier MSC Supplier No. 89465256
This tan burnishing floor pad is engineered for optimal performance with high-speed rotary disk floor machines exceeding 1500 rpm. It effectively cleans and polishes a variety of surfaces, including resilient tile, terrazzo, marble, and ceramic, consistently producing a "wet look" gloss in high-traffic environments.
  • Designed for rotary disk floor machines operating above 1500 rpm
  • Suitable for resilient tile, terrazzo, marble, and ceramic floors
  • Can be washed and reused for extended life
  • Corrosion-resistant materials ensure durability
  • Produces a high-gloss "wet look" finish
  • Features an 11/16 in thickness for consistent performance