Learn More
MSC AbilityOne Floor Pad, TopLine Speed Burnishing 3200 Series, 21 in Diameter
Catalog No. 502839595
Encompass
This burnishing floor pad, constructed with 3M floor pad materials, is designed for high-speed applications. With an 11/16 inch thickness, it is intended for rotary disk floor machines exceeding 1500 rpm and suitable for various floor types including resilient tile, terrazzo, marble, and ceramic.
  • Made of floor pad materials
  • 11/16 inch thick
  • Corrosion-resistant
  • Can be washed and reused
  • Restores gloss to dull floors in a single pass
Catalog No. 50-283-9595 Supplier MSC Supplier No. 89464655
This burnishing floor pad, constructed with 3M floor pad materials, is designed for high-speed applications. With an 11/16 inch thickness, it is intended for rotary disk floor machines exceeding 1500 rpm and suitable for various floor types including resilient tile, terrazzo, marble, and ceramic.
  • Made of floor pad materials
  • 11/16 inch thick
  • Corrosion-resistant
  • Can be washed and reused
  • Restores gloss to dull floors in a single pass