Learn More
Analytical Sales & Services Top PFA Sheet for 96 Position Radiochemistry Reactor
Catalog No. 5000806214
Encompass
Top PFA Sheet for 96 Position Radiochemistry Reactor\n\n• Used as “webbing” to hold vials in place and allow them to be moved via the transfer plate.\n• Lay PFA film down into the Transfer Plate and press your vials through.\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nNote: The PFA film/webbing is meant for one time use. Once vials are pushed through the PFA and then later removed, the PFA will be permanently stretched out and deformed.\n\nQty/Pkg: 10
Catalog No. 50-008-06214 Supplier Analytical Sales & Services Supplier No. 96957
Top PFA Sheet for 96 Position Radiochemistry Reactor\n\n• Used as “webbing” to hold vials in place and allow them to be moved via the transfer plate.\n• Lay PFA film down into the Transfer Plate and press your vials through.\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nNote: The PFA film/webbing is meant for one time use. Once vials are pushed through the PFA and then later removed, the PFA will be permanently stretched out and deformed.\n\nQty/Pkg: 10