Learn More
Perkin Elmer US LLC Top Plate Cover for Volatile Samples
Catalog No. 502112729
Encompass_Preferred
This cover is used with non-trough crystal plates and the Pike ATR accessories that are compatible with the Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-2729 Supplier Perkin Elmer US LLC Supplier No. L1272320
This cover is used with non-trough crystal plates and the Pike ATR accessories that are compatible with the Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.