Learn More
MSC Stanley Workmaster I-Beam Level, Aluminum, 48 in., 3 Vials, 1 ea.
Catalog No. 5000512823
Encompass
This 48 in. I-beam level features heavy-duty aluminum construction to provide rigidity and accuracy for various leveling applications. Designed for durability and precision, this tool includes three acrylic bubble vials and shock-absorbing end caps to protect against damage during heavy use.
  • Heavy-duty aluminum I-beam construction ensures durability and structural rigidity.
  • Features three vials for versatile measuring including one level and two plumb positions.
  • Accuracy rated at 0.0015 in./in.
  • Top-read center vial and high-visibility yellow vial covers for easy readability.
  • Shock-absorbing end caps prevent denting and withstand heavy site use.
  • Integrated hang holes allow for convenient storage.
Catalog No. 50-005-12823 Supplier MSC Supplier No. 06530687
This 48 in. I-beam level features heavy-duty aluminum construction to provide rigidity and accuracy for various leveling applications. Designed for durability and precision, this tool includes three acrylic bubble vials and shock-absorbing end caps to protect against damage during heavy use.
  • Heavy-duty aluminum I-beam construction ensures durability and structural rigidity.
  • Features three vials for versatile measuring including one level and two plumb positions.
  • Accuracy rated at 0.0015 in./in.
  • Top-read center vial and high-visibility yellow vial covers for easy readability.
  • Shock-absorbing end caps prevent denting and withstand heavy site use.
  • Integrated hang holes allow for convenient storage.