Learn More
Cole-Parmer Cole-Parmer Top Seal Screw Thread Closure, Clear, Molded 10 mil Septa, 8-425 mm; 100/pk
Catalog No. NC1568744 Shop All Cole-Parmer Products
Encompass_Preferred
Select from a variety of caps, septa and inserts that fit your application and needs.This screw thread cap and septa is the closure only; vial sold separately.
Catalog No. NC1568744 Supplier Cole-Parmer Supplier No. 9872110
Select from a variety of caps, septa and inserts that fit your application and needs.This screw thread cap and septa is the closure only; vial sold separately.