Learn More
Analytical Sales & Services Top Silicone Mat for Nano Nest
Catalog No. 5000804998
Encompass
Top Silicone Mat for Nano Nest\n\n• Compatible with all Nano Nest Parallel Synthesis and Photoredox Blocks
Catalog No. 50-008-04998 Supplier Analytical Sales & Services Supplier No. 1626005
Add to Cart
Add to Cart
Top Silicone Mat for Nano Nest\n\n• Compatible with all Nano Nest Parallel Synthesis and Photoredox Blocks