Learn More
Hach Company Ceramic Top Magnetic Stirrer, 4x4 in., 115 Vac
Supplier:  Hach Company 2881200
Encompass
Cimarec Ceramic top stirrer features a reflective, white ceramic that is seamlesseasy to clean, and resistant to alkali/acid solutions. Low profile increases efficiencyand reduces spill risk. StirTrac™ technology offers improved slow speed stirring, consistent speed control, adjustable speed range from 50-1200 RPM, and rapid braking. Detachable cord. Integral ringstand/support rod holder (order rod separately).
Catalog No. 50-199-6760