Learn More
Anton Paar Top-tier model with customizable methods, including ASTM D6226 for open-cell foam. PowderProtect prevents contamination. Built-in Peltier control (3–60 °C) removes need for an external bath. Accurate results for 4.5–135 cm³ samples.
Catalog No. 5000580475
Encompass_Preferred
Performance specificationsAccuracy: 0.015 %Number reference volumes: 2 (TruPyc)Nominal reference volumes: 50 cm3 & 8 cm3Inserts available: 135 cm3, 50 cm3, 10 cm3, 4.5 cm3, 1.8 cm3, 0.25 cm3Preparation modes: Flow, pulse, vauum, noneGas expansion direction: Bi-directional flow (PowderProtect)Built-in temperature control: 3 °C to 60 °CTemperature stability: 0.02 °CBuilt-in & customizable method librarySample chamber closure: Repeatable dual-turn self-aligning lid (TruLock)Display & control:7-inch TFT WVGA (800×480) PCAP touchscreenGraphical user interfaceTransducer accuracy: Better than 0.1 %Pressure reading resolution: 0.00001 psiDigital pressure readout: 0.000001 bar
Catalog No. 50-005-80475 Supplier Anton Paar Supplier No. 387703
Please contact your sales representative for a quote or further ordering instructions.
Performance specificationsAccuracy: 0.015 %Number reference volumes: 2 (TruPyc)Nominal reference volumes: 50 cm3 & 8 cm3Inserts available: 135 cm3, 50 cm3, 10 cm3, 4.5 cm3, 1.8 cm3, 0.25 cm3Preparation modes: Flow, pulse, vauum, noneGas expansion direction: Bi-directional flow (PowderProtect)Built-in temperature control: 3 °C to 60 °CTemperature stability: 0.02 °CBuilt-in & customizable method librarySample chamber closure: Repeatable dual-turn self-aligning lid (TruLock)Display & control:7-inch TFT WVGA (800×480) PCAP touchscreenGraphical user interfaceTransducer accuracy: Better than 0.1 %Pressure reading resolution: 0.00001 psiDigital pressure readout: 0.000001 bar