Learn More
Anton Paar Top-tier model for small volumes with customizable methods. Accurate results from 0.1–10 cm³. PowderProtect prevents contamination. Built-in Peltier control (3–60 °C) eliminates external bath. Ideal for R&D and limited samples.
Catalog No. 5000580483
Encompass_Preferred
Performance specificationsAccuracy: 0.075 %Number reference volumes: 2 (TruPyc)Nominal reference volumes: 2 cm3 & 5 cm3Inserts available: 10 cm3, 4.5 cm3, 1.8 cm3, 0.25 cm3, 0.1 cm3Preparation modes: Flow, pulse, vauum, noneGas expansion direction: Bi-directional flow (PowderProtect)Built-in temperature control: 3 °C to 60 °CTemperature stability: 0.02 °CBuilt-in & customizable method librarySample chamber closure: Repeatable dual-turn self-aligning lid (TruLock)Display & control:7-inch TFT WVGA (800×480) PCAP touchscreenGraphical user interfaceTransducer accuracy: Better than 0.1 %Pressure reading resolution: 0.00001 psiDigital pressure readout: 0.000001 bar
Catalog No. 50-005-80483 Supplier Anton Paar Supplier No. 387704
Please contact your sales representative for a quote or further ordering instructions.
Performance specificationsAccuracy: 0.075 %Number reference volumes: 2 (TruPyc)Nominal reference volumes: 2 cm3 & 5 cm3Inserts available: 10 cm3, 4.5 cm3, 1.8 cm3, 0.25 cm3, 0.1 cm3Preparation modes: Flow, pulse, vauum, noneGas expansion direction: Bi-directional flow (PowderProtect)Built-in temperature control: 3 °C to 60 °CTemperature stability: 0.02 °CBuilt-in & customizable method librarySample chamber closure: Repeatable dual-turn self-aligning lid (TruLock)Display & control:7-inch TFT WVGA (800×480) PCAP touchscreenGraphical user interfaceTransducer accuracy: Better than 0.1 %Pressure reading resolution: 0.00001 psiDigital pressure readout: 0.000001 bar