Learn More
Aviva Systems Biology Corp TOP1 Antibody - N-terminal region (ARP61035_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOP1. It was validated on Western Blot. . Alternate names: TOPIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-236