Learn More
MyBioSource Mouse ELISA Kit, DNA Topoisomerase 1 Autoantibody (Anti-TOP1)
SDP
Supplier:  MyBioSource MBS991563996STRIPWELLS
Encompass_Preferred
The MyBioSource Mouse ELISA Kit quantitatively measures Anti-TOP1 in various mouse biological fluids using a specific antibody-coated 96-well plate. After binding and washing steps, a biotinylated detection antibody and HRP conjugate are added. Substrate reaction reveals Anti-TOP1 levels by color intensity, read at 450 nm.
Catalog No. NC2685008