Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7153MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: TOP2A/1361. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HeLa cells or Tonsil. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Topoisomerase II alpha fragment (aa1352-1493). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 170kDa.Chromosome: 17q21.2. Gene Symbol: TOP2A. Gene ID: 7153. Ensemble: ENSG00000131747. Uniprot: XP_011523467.
Catalog No. 50-188-5233