Learn More
Aviva Systems Biology Corp TOP2B Antibody - middle region (ARP33079_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33079_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOP2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TOP2B. Alternate names: TOPIIB, top2betaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54876