Learn More
Medchemexpress LLC Topanol CA | 1843-03-4 | MFCD00072238 | 98.1% | 544.81 g/mol | C37H52O3 | 5 MG
SDP
Catalog No. 5000264280
Encompass_Preferred
Topanol CA is a phenolic antioxidant used to protect polymer resins and light-colored rubber products from oxidative degradation. Supplied as a white to off-white solid with high purity, it functions as a stabilizer during compounding and processing to improve color stability and service life.
  • Acts as a phenolic antioxidant and stabilizer for polymers.
  • Helps maintain color and reduce discoloration in light-colored rubber.
  • High purity solid, suitable for laboratory and formulation use.
  • Effective in resins such as polypropylene, polyethylene, ABS, and PVC.
  • Documentation available: safety data sheet and certificate of analysis.
Catalog No. 50-002-64280 Supplier Medchemexpress LLC Supplier No. HY1127945MG
Topanol CA is a phenolic antioxidant used to protect polymer resins and light-colored rubber products from oxidative degradation. Supplied as a white to off-white solid with high purity, it functions as a stabilizer during compounding and processing to improve color stability and service life.
  • Acts as a phenolic antioxidant and stabilizer for polymers.
  • Helps maintain color and reduce discoloration in light-colored rubber.
  • High purity solid, suitable for laboratory and formulation use.
  • Effective in resins such as polypropylene, polyethylene, ABS, and PVC.
  • Documentation available: safety data sheet and certificate of analysis.