Learn More
Grainger Topical Antiseptic 128oz Bottle
Catalog No. 502749878
Encompass_Preferred
Cumberland Swan Topical Antiseptic Liquid is an antiseptic solution for first aid and personal care use. It contains 0.13% benzalkonium chloride as the active ingredient. This clear, colorless liquid can be used to cleanse and deodorize skin, minor wounds, and skin irritations. It also helps prevent infection in minor cuts, scrapes, and burns. This 8 oz bottle of antiseptic liquid is made in the USA. It can be used by the entire family.
Catalog No. 50-274-9878 Supplier Grainger Supplier No. 14U290
Cumberland Swan Topical Antiseptic Liquid is an antiseptic solution for first aid and personal care use. It contains 0.13% benzalkonium chloride as the active ingredient. This clear, colorless liquid can be used to cleanse and deodorize skin, minor wounds, and skin irritations. It also helps prevent infection in minor cuts, scrapes, and burns. This 8 oz bottle of antiseptic liquid is made in the USA. It can be used by the entire family.