Learn More
TARGETMOL CHEMICALS INC Topiramate 200MG
SDP
Catalog No. 502880694
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Topiramate (RWJ 17021) is a unique antiseizure medication that is used in the treatment of partial and generalized seizures. Topiramate has been rarely associated with hepatic injury and largely when used in combination with other anticonvulsant medications. Purity 98%
Catalog No. 50-288-0694 Supplier TARGETMOL CHEMICALS INC Supplier No. T0675200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Topiramate (RWJ 17021) is a unique antiseizure medication that is used in the treatment of partial and generalized seizures. Topiramate has been rarely associated with hepatic injury and largely when used in combination with other anticonvulsant medications. Purity 98%