Learn More
Selleck Chemical LLC Topiramate S1438-1g
SDP
Catalog No. 503110233
Encompass_Preferred
Topiramate (MCN 4853 RWJ 17021) is a mutil-targeted inhibitor including voltage-gated sodium channel and calcium channel AMPA/kainate receptor and carbonic anhydrase used to treat epilepsy
Catalog No. 50-311-0233 Supplier Selleck Chemical LLC Supplier No. S14381G
Topiramate (MCN 4853 RWJ 17021) is a mutil-targeted inhibitor including voltage-gated sodium channel and calcium channel AMPA/kainate receptor and carbonic anhydrase used to treat epilepsy