Learn More
Medchemexpress LLC Topiramate (McN 4853) | 97240-79-4 | 99.8% | 500 MG
SDP
Catalog No. 5000450936
Encompass_Preferred
Topiramate is a broad-spectrum antiepileptic agent and a GluR5 receptor antagonist. It produces its antiepileptic effects by enhancing GABAergic activity, inhibiting kainate/AMPA receptors, inhibiting voltage-sensitive sodium and calcium channels, increasing potassium conductance, and inhibiting carbonic anhydrase. This product is for research use only.
  • Broad-spectrum antiepileptic agent
  • GluR5 receptor antagonist
  • Enhances GABAergic activity
  • Inhibits kainate/AMPA receptors
  • Inhibits voltage-sensitive sodium and calcium channels
  • Increases potassium conductance
  • Inhibits carbonic anhydrase
Catalog No. 50-004-50936 Supplier Medchemexpress LLC Supplier No. HYB0122500MG
Topiramate is a broad-spectrum antiepileptic agent and a GluR5 receptor antagonist. It produces its antiepileptic effects by enhancing GABAergic activity, inhibiting kainate/AMPA receptors, inhibiting voltage-sensitive sodium and calcium channels, increasing potassium conductance, and inhibiting carbonic anhydrase. This product is for research use only.
  • Broad-spectrum antiepileptic agent
  • GluR5 receptor antagonist
  • Enhances GABAergic activity
  • Inhibits kainate/AMPA receptors
  • Inhibits voltage-sensitive sodium and calcium channels
  • Increases potassium conductance
  • Inhibits carbonic anhydrase