Learn More
Medchemexpress LLC Topiramate (Standard) | 97240-79-4 | 99.8% | 10 MM * 1 ML
SDP
Catalog No. 5000450752
Encompass_Preferred
Topiramate is a broad-spectrum antiepileptic agent and a GluR5 receptor antagonist. Its antiepileptic effects are attributed to enhancing GABAergic activity, inhibiting kainate/AMPA receptors, blocking voltage-sensitive sodium and calcium channels, increasing potassium conductance, and inhibiting carbonic anhydrase.
  • Enhances GABAergic activity
  • Inhibits kainate/AMPA receptors
  • Blocks voltage-sensitive sodium and calcium channels
  • Increases potassium conductance
  • Inhibits carbonic anhydrase
  • Use-dependent inhibition of voltage-dependent Na+ channels in neurons
  • Potentiation of GABA (γ-amino-butyric acid)-induced Cl- influx
  • Antagonizes kainate/AMPA receptors for inhibitory effects on inward currents
Catalog No. 50-004-50752 Supplier Medchemexpress LLC Supplier No. HYB012210MM/1ML
Topiramate is a broad-spectrum antiepileptic agent and a GluR5 receptor antagonist. Its antiepileptic effects are attributed to enhancing GABAergic activity, inhibiting kainate/AMPA receptors, blocking voltage-sensitive sodium and calcium channels, increasing potassium conductance, and inhibiting carbonic anhydrase.
  • Enhances GABAergic activity
  • Inhibits kainate/AMPA receptors
  • Blocks voltage-sensitive sodium and calcium channels
  • Increases potassium conductance
  • Inhibits carbonic anhydrase
  • Use-dependent inhibition of voltage-dependent Na+ channels in neurons
  • Potentiation of GABA (γ-amino-butyric acid)-induced Cl- influx
  • Antagonizes kainate/AMPA receptors for inhibitory effects on inward currents