Learn More
Medchemexpress LLC Topiramate (Standard) | 97240-79-4 | MFCD00865320 | 25 MG
SDP
Catalog No. 5000406814
Encompass_Preferred
Topiramate (Standard) is a hexose derivative that acts as an anticonvulsant and sodium channel blocker. It is primarily used as a laboratory chemical and for the manufacture of substances, with applications in antiepileptic treatment and migraine prevention. This compound is supplied as a solid and maintains stability under recommended storage conditions.
  • Functions as an anticonvulsant and sodium channel blocker
  • Used in laboratory chemicals and for substance manufacturing
  • Recommended storage at 4°C, sealed, away from light and moisture
  • Stable under recommended storage conditions
  • Solid appearance
  • For research use only
Catalog No. 50-004-06814 Supplier Medchemexpress LLC Supplier No. 25MGHYB0122R
Topiramate (Standard) is a hexose derivative that acts as an anticonvulsant and sodium channel blocker. It is primarily used as a laboratory chemical and for the manufacture of substances, with applications in antiepileptic treatment and migraine prevention. This compound is supplied as a solid and maintains stability under recommended storage conditions.
  • Functions as an anticonvulsant and sodium channel blocker
  • Used in laboratory chemicals and for substance manufacturing
  • Recommended storage at 4°C, sealed, away from light and moisture
  • Stable under recommended storage conditions
  • Solid appearance
  • For research use only