Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Topiramate (standard) | 97240-79-4 | MFCD00865320 | 99.8% | 10 MG
SDP

Catalog No. 5000298230
Encompass_Preferred

Topiramate analytical standard supplied as a small, weighed quantity for analytical and research use. Intended for preparation of calibration standards, method development, and laboratory QC; not for human therapeutic use.

  • Analytical reference standard for topiramate.
  • Packaged as a weighed 10 mg unit for solution preparation.
  • High purity suitable for analytical work (≈99.8%).
  • Molecular formula C12H21NO8S; molecular weight 339.36 g/mol.
  • Solubility notes: DMSO ≥100 mg/mL; water ≈4 mg/mL (may require ultrasonic).
  • For research use only; handle according to the safety data sheet.

Catalog No. 50-002-98230 Supplier Medchemexpress LLC Supplier No. HYB0122R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Topiramate analytical standard supplied as a small, weighed quantity for analytical and research use. Intended for preparation of calibration standards, method development, and laboratory QC; not for human therapeutic use.

  • Analytical reference standard for topiramate.
  • Packaged as a weighed 10 mg unit for solution preparation.
  • High purity suitable for analytical work (≈99.8%).
  • Molecular formula C12H21NO8S; molecular weight 339.36 g/mol.
  • Solubility notes: DMSO ≥100 mg/mL; water ≈4 mg/mL (may require ultrasonic).
  • For research use only; handle according to the safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.