Learn More
Medchemexpress LLC HY-14874 25mg Medchemexpress, Topiroxostat CAS:577778-58-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1487425MG
Encompass_Preferred
Medchemexpress, HY-14874 25mg Topiroxostat CAS:577778-58-6 Topiroxostat (FYX-051) is a potent and orally active xanthine oxidoreductase (XOR) inhibitor with an IC50 value of 5.3 nM and a Ki value of 5.7 nM. Topiroxostat exhibits weak CYP3A4-inhibitory activity (18.6%). Topiroxostat has the potential for hyperuricemia treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9921