Learn More
MSC SKILCRAFT Floor Pad, TopLine Pre-Burnishing 5000 Series, 18 inch Diameter
Catalog No. 502839609
Encompass
This TopLine pre-burnishing floor pad is designed for light floor scrubbing with floor machines prior to burnishing. It serves as an excellent daily cleaning pad, offering minimal finish removal. Its open construction resists clogging and allows for easy cleaning.
  • Designed to prepare finished floors for optimal gloss.
  • Provides light scrubbing to remove surface-level dirt, scuff marks, and other imperfections.
  • Formulated for minimal finish removal.
  • Effective at lifting surface-level soil.
  • Effective at lifting surface-level soil.
  • Excellent choice for daily cleaning.
Catalog No. 50-283-9609 Supplier MSC Supplier No. 89465769
This TopLine pre-burnishing floor pad is designed for light floor scrubbing with floor machines prior to burnishing. It serves as an excellent daily cleaning pad, offering minimal finish removal. Its open construction resists clogging and allows for easy cleaning.
  • Designed to prepare finished floors for optimal gloss.
  • Provides light scrubbing to remove surface-level dirt, scuff marks, and other imperfections.
  • Formulated for minimal finish removal.
  • Effective at lifting surface-level soil.
  • Effective at lifting surface-level soil.
  • Excellent choice for daily cleaning.