Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, without competent cells

Catalog No. 452640
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Reactions
25 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-264-0 10 Reactions
45-064-0 25 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-264-0 Supplier Invitrogen™ Supplier No. 452640
  • $353.00
    $328.65 / Each of 1

    Save $24.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Topoisomerase I-activated pCR™II-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water

Includes the pCR™II-TOPO TA vector with dual T7 and SP6 promoters. By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable cloning method, featuring a 5-minute cloning reaction and 3-step protocol, and yielding up to 95% recombinants.

Includes the pCR™II-TOPO TA vector with dual T7 and SP6 promoters. By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable cloning method, featuring a 5-minute cloning reaction and 3-step protocol, and yielding up to 95% recombinants. Resistant to Kanamycin (KanR) andZeocin™ (ZeoR)
Kit Options: TOPO TA Cloning Dual Promoter Kits
TOPO TA Cloning Dual Promoter kits can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:

  • General cloning: TOP10 cells (Cat. Nos. K4600-J10, K4600-01, K4600-40)
  • High-efficiency cloning: TOP10 Electrocomp™ cells (Cat. Nos. K4660-01, K4660-40)
  • General cloning, bacteriophage T1 resistance: DH5α-T1R cells (Cat. Nos. K4620-01, K4620-40)
  • Fast growth: Mach1™ -T1R chemically competent E. coli (Cat. No. K4610-20)
  • Repressor/induction needs: TOP10F' cells (Cat. Nos. K4650-01, K4650-40)
  • Provide your own cells (Cat. Nos. 450640 and 452640)

  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and SP6 promoters for efficient in vitro transcription
  • M13 forward and reverse primer sites for sequencing
  • 16 convenient restriction sites, including EcoRI, flanking your insert for subsequent excision or subcloning
  • Kanamycin and ampicillin resistance for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR), Kanamycin (KanR)
Cloning Method TOPO™-TA
Content And Storage Box 1:
• Topoisomerase I-activated pCR™II-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.
Format Kit
For Use With (Application) Cloning and subsequent in vitro transcription
Includes Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water
Product Line TOPO
Product Type TA Cloning Kit
Promoter T7, SP6
Quantity 10 Reactions
Vector TOPO-TA Vectors
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.