Learn More
TOPOGEN INC Topoisomerase I Assay Kit 500 Assays Includes Top1
Catalog No. NC1818637
Encompass
This kit contains reagents for routine detection of type I topoisomerase activity. DNA markers (included) allow unequivocal detection of the enzyme. The assay is specific for topoisomerase I; (eukaryotic type II topoisomerases require MgCl2 and ATP). The standard topo I assay kit is supplied with EDTA containing reaction buffer (minus Mg++) in order to prevent nucleolytic nicking of the supercoiled substrate (although we note that some eukaryotic type I activities are stimulated by Mg++). This assay will work even with crude extracts.
Catalog No. NC1818637 Supplier TOPOGEN INC Supplier No. TG10153A
This kit contains reagents for routine detection of type I topoisomerase activity. DNA markers (included) allow unequivocal detection of the enzyme. The assay is specific for topoisomerase I; (eukaryotic type II topoisomerases require MgCl2 and ATP). The standard topo I assay kit is supplied with EDTA containing reaction buffer (minus Mg++) in order to prevent nucleolytic nicking of the supercoiled substrate (although we note that some eukaryotic type I activities are stimulated by Mg++). This assay will work even with crude extracts.