Learn More
New England Biolabs, Inc. Topoisomerase I (E. coli) – 100 units
SDP
Catalog No. 50811724
Encompass
Topoisomerase I (E. coli) catalyzes the relaxation of negatively supercoiled DNA. Topoisomerase I has also been implicated in knotting and unknotting DNA and in linking complementary rings of single-stranded DNA into double-stranded rings. The intact holoenzyme is a 97 kDa protein.
Catalog No. 50-811-724 Supplier New England Biolabs, Inc. Supplier No. M0301S
Topoisomerase I (E. coli) catalyzes the relaxation of negatively supercoiled DNA. Topoisomerase I has also been implicated in knotting and unknotting DNA and in linking complementary rings of single-stranded DNA into double-stranded rings. The intact holoenzyme is a 97 kDa protein.