Learn More
Sigma Aldrich Fine Chemicals Biosciences Topoisomerase II alpha hum
Catalog No. 502905504
Encompass
Topoisomerase II is encoded by TOP2A gene which is located on 17q21.2 in the human chromosome. It is essential for controlling the topological state of DNA by reversibly breaking the DNA strands during replication recombination and transcription. TOP2A expression increases at late S phase and during G2/M of the cell cycle and decreases after mitosis. Polymorphism with TOP2A causes chromosome instability and tumorigenesis. TOP2A causes tumor proliferation and invasiveness in various cancer types like breast cancer ovarian cancer and prostate cancer.
Catalog No. 50-290-5504 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T8944100UN
Topoisomerase II is encoded by TOP2A gene which is located on 17q21.2 in the human chromosome. It is essential for controlling the topological state of DNA by reversibly breaking the DNA strands during replication recombination and transcription. TOP2A expression increases at late S phase and during G2/M of the cell cycle and decreases after mitosis. Polymorphism with TOP2A causes chromosome instability and tumorigenesis. TOP2A causes tumor proliferation and invasiveness in various cancer types like breast cancer ovarian cancer and prostate cancer.