Learn More
ABclonal Technology Topoisomerase II alpha/TOP2A
SDP
Supplier:  ABclonal Technology A2649720UL
Encompass_Preferred
This gene encodes a DNA topoisomerase an enzyme that controls and alters the topologic states of DNA during transcription This nuclear enzyme is involved in processes such as chromosome condensation chromatid separation and the relief of torsional stress that occurs during DNA transcription and replication It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another thus altering the topology of DNA Two forms of this enzyme exist as likely products of a gene duplication event The gene encoding this form alpha is localized to chromosome 17 and the beta gene is localized to chromosome 3 The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance Reduced activity of this enzyme may also play a role in ataxia-telangiectasia
Catalog No. 50-000-11416