Learn More
Medchemexpress LLC Topoisomerase IIα-in-8 | 1251578-67-2 | 314.32 | C15H15FN6O | 1 MG
SDP
Catalog No. 5000301573
Encompass_Preferred
Topoisomerase IIα-IN-8 is a research-grade small-molecule inhibitor of human DNA topoisomerase IIα reported as a weak inhibitor (IC50 = 462 ± 38.0 μM). It is supplied as a powder for laboratory use, has molecular formula C15H15FN6O and molecular weight 314.32 g·mol-1, and is provided with downloadable product and safety documentation. For research use only.
  • Weak inhibitor of human DNA topoisomerase IIα (IC50 = 462 ± 38.0 μM).
  • Shows limited antiproliferative activity (GI50 > 100 μM in HEK293 and HT-29 cells).
  • Supplied as a powder with specified storage recommendations for powder and solutions.
  • Molecular formula C15H15FN6O; molecular weight 314.32 g·mol-1.
  • Datasheet and safety data sheet available for handling and purity details.
Catalog No. 50-003-01573 Supplier Medchemexpress LLC Supplier No. HYW0313071MG
Topoisomerase IIα-IN-8 is a research-grade small-molecule inhibitor of human DNA topoisomerase IIα reported as a weak inhibitor (IC50 = 462 ± 38.0 μM). It is supplied as a powder for laboratory use, has molecular formula C15H15FN6O and molecular weight 314.32 g·mol-1, and is provided with downloadable product and safety documentation. For research use only.
  • Weak inhibitor of human DNA topoisomerase IIα (IC50 = 462 ± 38.0 μM).
  • Shows limited antiproliferative activity (GI50 > 100 μM in HEK293 and HT-29 cells).
  • Supplied as a powder with specified storage recommendations for powder and solutions.
  • Molecular formula C15H15FN6O; molecular weight 314.32 g·mol-1.
  • Datasheet and safety data sheet available for handling and purity details.