Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Topoisomerase II inhibitor 14 | 305343-00-4 | MFCD06592467 | 99.7% | 332.19 g/mol | C15H11Cl2N5 | 5 MG
SDP

Catalog No. 5000282465
Encompass_Preferred

Topoisomerase II inhibitor 14 is a research-grade small molecule inhibitor of topoisomerase II. It is described as a potent inhibitor that induces apoptosis, arrests the cell cycle at S and G2-M phases, and exhibits antioxidant activity. The compound is provided as a high-purity solid and is intended for laboratory research use only.

  • Potent topoisomerase II inhibition with cellular activity.
  • Induces apoptosis and cell cycle arrest for mechanistic studies.
  • Exhibits antioxidant effects useful in oxidative stress research.
  • High purity appropriate for biochemical and cellular assays.
  • Available in small packs suitable for screening and pilot studies.
  • Soluble in dimethyl sulfoxide (DMSO) with recommended warming and sonication.
  • Storage conditions specified for solid and solution to preserve stability.

Catalog No. 50-002-82465 Supplier Medchemexpress LLC Supplier No. HYW2785825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Topoisomerase II inhibitor 14 is a research-grade small molecule inhibitor of topoisomerase II. It is described as a potent inhibitor that induces apoptosis, arrests the cell cycle at S and G2-M phases, and exhibits antioxidant activity. The compound is provided as a high-purity solid and is intended for laboratory research use only.

  • Potent topoisomerase II inhibition with cellular activity.
  • Induces apoptosis and cell cycle arrest for mechanistic studies.
  • Exhibits antioxidant effects useful in oxidative stress research.
  • High purity appropriate for biochemical and cellular assays.
  • Available in small packs suitable for screening and pilot studies.
  • Soluble in dimethyl sulfoxide (DMSO) with recommended warming and sonication.
  • Storage conditions specified for solid and solution to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.