Learn More
Selleck Chemical LLC Topotecan S9321-1G
SDP
Catalog No. 5000023704
Encompass_Preferred
Topotecan(NSC609699 Nogitecan SKFS 104864A) is an antineoplastic agent used to treat ovarian cancer that works by inhibiting DNA topoisomerases
Catalog No. 50-000-23704 Supplier Selleck Chemical LLC Supplier No. S93211G
Topotecan(NSC609699 Nogitecan SKFS 104864A) is an antineoplastic agent used to treat ovarian cancer that works by inhibiting DNA topoisomerases