Learn More
Selleck Chemical LLC Topotecan HCl 50mg 119413-54-6 NSC609699, Nogitecan HCl, SKFS 104864A
SDP
Catalog No. 507115
Encompass_Preferred
Topotecan HCl (NSC609699, Nogitecan, SKFS 104864A) is a topoisomerase I inhibitor for MCF-7 Luc cells and DU-145 Luc cells with IC50 of 13 nM and 2 nM in cell-free assays, respectively. Topotecan HCl induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-711-5 Supplier Selleck Chemical LLC Supplier No. S123150MG
Topotecan HCl (NSC609699, Nogitecan, SKFS 104864A) is a topoisomerase I inhibitor for MCF-7 Luc cells and DU-145 Luc cells with IC50 of 13 nM and 2 nM in cell-free assays, respectively. Topotecan HCl induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.