Learn More
Medchemexpress LLC Topotecan hydrochloride | 119413-54-6 | 98.1% | 457.91 | 1 ML
SDP
Catalog No. 5000349611
Encompass_Preferred
Topotecan hydrochloride is a Topoisomerase I inhibitor with potent antineoplastic activities, intended for research use only.
  • Inhibits Topoisomerase I
  • Displays potent antineoplastic activities
  • Inhibits proliferation of human glioma cells and glioma stem cells
  • Induces cell cycle arrest in G0/G1 and S phases
  • Promotes apoptosis
Catalog No. 50-003-49611 Supplier Medchemexpress LLC Supplier No. HY13768A10MM/1ML
Topotecan hydrochloride is a Topoisomerase I inhibitor with potent antineoplastic activities, intended for research use only.
  • Inhibits Topoisomerase I
  • Displays potent antineoplastic activities
  • Inhibits proliferation of human glioma cells and glioma stem cells
  • Induces cell cycle arrest in G0/G1 and S phases
  • Promotes apoptosis