Learn More
Medchemexpress LLC Topotecan hydrochloride | 119413-54-6 | MFCD00866235 | 99.9% | 457.91 | 100 MG
SDP
Catalog No. 5000350578
Encompass_Preferred
Topotecan hydrochloride is a potent Topoisomerase I inhibitor with significant antineoplastic activities. It is intended for research and analytical applications, playing a role in studies related to cancer, autophagy, and apoptosis.
  • Potent Topoisomerase I inhibitor
  • Demonstrates antineoplastic activities
  • Inhibits the proliferation of human glioma cells
  • Induces cell cycle arrest in G0/G1 and S phases
  • Promotes apoptosis in various cell lines
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-003-50578 Supplier Medchemexpress LLC Supplier No. HY13768A100MG
Topotecan hydrochloride is a potent Topoisomerase I inhibitor with significant antineoplastic activities. It is intended for research and analytical applications, playing a role in studies related to cancer, autophagy, and apoptosis.
  • Potent Topoisomerase I inhibitor
  • Demonstrates antineoplastic activities
  • Inhibits the proliferation of human glioma cells
  • Induces cell cycle arrest in G0/G1 and S phases
  • Promotes apoptosis in various cell lines
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 2 years or -20°C for 1 year