Learn More
Medchemexpress LLC Topotecan hydrochloride | 119413-54-6 | MFCD00866235 | 99.9% | 457.91 g/mol | C23H24ClN3O5 | 5 MG
SDP
Catalog No. 5000222876
Encompass_Preferred
Topotecan hydrochloride is the hydrochloride salt of topotecan, a potent topoisomerase I inhibitor used in oncology research. It induces DNA damage and apoptosis in tumor cells and is supplied for research use only. Typical solvent compatibility includes DMSO.
  • Potent topoisomerase I inhibitor with antineoplastic activity.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for cellular and molecular studies.
  • Soluble in DMSO for assay and dosing preparations.
  • Used to induce DNA damage, cell cycle arrest, and apoptosis in tumor models.
  • Intended for research use only; not for human or diagnostic applications.
Catalog No. 50-002-22876 Supplier Medchemexpress LLC Supplier No. HY13768A5MG
Topotecan hydrochloride is the hydrochloride salt of topotecan, a potent topoisomerase I inhibitor used in oncology research. It induces DNA damage and apoptosis in tumor cells and is supplied for research use only. Typical solvent compatibility includes DMSO.
  • Potent topoisomerase I inhibitor with antineoplastic activity.
  • Hydrochloride salt form for improved stability and handling.
  • High purity suitable for cellular and molecular studies.
  • Soluble in DMSO for assay and dosing preparations.
  • Used to induce DNA damage, cell cycle arrest, and apoptosis in tumor models.
  • Intended for research use only; not for human or diagnostic applications.