Learn More
Cayman Chemical Topotecan (hydrochloride), 119413-54-6, 50 mg
SDP
Supplier:  Cayman Chemical 1412950
Encompass_Preferred
A potent inhibitor of DNA topoisomerase I that induces cytoxicity and DNA damage in human HT-29 colon adenocarcinoma cells (IC50s = 33 nM and 0.28 μM, respectively; demonstrates a broad spectrum of antitumor activity against a variety of other tumors including ovarian cancer, cervical cancer, neuroblastoma, brainstem glioma, and Ewing’s sarcoma
Catalog No. NC1406706