Learn More
Enzo Life Sciences Topotecan . hydrochloride (5mg). CAS: 119413-54-6
SDP
Supplier:  Enzo Life Sciences ALX350133M005
Encompass_Preferred
Potent antitumor agent. Inhibitor of DNA-topoisomerase. Analog of (S)-(+)-camptothecin (ALX-350-015). Used in chemotherapy of several different cancers. Alternative name: 9-[(Dimethylamino)methyl]-10-hydroxy-(20S)-camptothecin . HCl. Purity: ≥98% (HPLC). Appearance: Light yellow to greenish powder. Solubility: Soluble in water (1mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8751