Learn More
Abcam Topotecan hydrochloride (SKF 104864), Topoisomerase inhibitor, 10MG
Catalog No. 502707625
Encompass_Preferred
MW 457.9 Da, Purity >98%. Topoisomerase I inhibitor. Water-soluble derivative of camptothecin (ab120115). Anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7625 Supplier Abcam Supplier No. AB12038810MG
Add to Cart
Add to Cart
MW 457.9 Da, Purity >98%. Topoisomerase I inhibitor. Water-soluble derivative of camptothecin (ab120115). Anticancer agent.
The product is subject to the following: Abcam Restricted Use Statement