Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte Topsoil Tour, Digital Version
SDPSCI_ED

Catalog No. S89508 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
4 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89508 4 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89508 Supplier LaMotte Supplier No. 5973
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Get the dirt on soil! A classroom investigation into the physical and chemical properties of soil.

  • Each student completes seven units while conducting his/her own soil tests for texture, pH, nitrogen, phosphorus and potassium
  • TesTabs reagent systems
  • Instructor Manual with full details on setting up and answer key
  • Student Manual with test procedures, data sheets and puzzles
  • Students work in groups and individually
  • Works on tablets or computers, or print paper copies for distribution
  • Great for inclusion classrooms.

Specifications

Grade 4 to 8
Age 9 to 14
For Use With (Application) Investigating The Physical and Chemical Properties of Soil
Includes Instructor Manual, Student Manual with Test Procedures, Data Sheets and Puzzles
Requires Tablets or Computers, or Printer
Class Size 1 Class
Product Type Mini Classroom Curriculum
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.