Learn More
Aviva Systems Biology Corp TOR1A Antibody - C-terminal region (ARP63400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOR1A. It was validated on Western Blot. . Alternate names: DQ2, DYT1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-959